N,N-Dimethylformamide ≥98%, TECHNICAL
Supplier: VWR Chemicals
About this item
Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
- CAS Number:68-12-2
- Formula:HCON(CH₃)₂
- MDL Number:MFCD00003284
- Molecular Weight:73.09 g/mol
- Merck Index:13,03269
- Boiling Pt:153 °C (1013 hPa)
- Melting Pt:–60,5 °C
- Density:0.949 g/cm³ (20 °C)
- Flash Pt:57.5 °C (DIN 51755) (closed cup)
- Storage Temperature:Ambient
- EINECS:200-679-5
- UN:2265
- ADR:3,III
- REACH:01-2119475605-32
3 Options Available Below
Product Details & Documents
Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
- CAS Number:68-12-2
- Formula:HCON(CH₃)₂
- MDL Number:MFCD00003284
- Molecular Weight:73.09 g/mol
- Merck Index:13,03269
- Boiling Pt:153 °C (1013 hPa)
- Melting Pt:–60,5 °C
- Density:0.949 g/cm³ (20 °C)
- Flash Pt:57.5 °C (DIN 51755) (closed cup)
- Storage Temperature:Ambient
- EINECS:200-679-5
- UN:2265
- ADR:3,III
- REACH:01-2119475605-32
Dokumente
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Verpackungsinformationen
27442969.pdf P20_VWRC23469.298_SPEC_EN.pdf P20_VWRC23469.367_SPEC_EN.pdf spec_VWRC23469.298 spec_VWRC23469.298
Produktspezifikation
More
SDS_EN_UK_VWRC23469.pdf sds_VWRC23469
Sicherheitsdatenblatt
VWR® Green Solvents for Sustainable Chemistry.pdf
Produktliteratur
Specifications for Product Family
Specification Test Results
- Assay:Min. 98 %
- Refractive index (20°C):1.429 - 1.431
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



