Order Entry
Germany
Orders LinkContactUsLinkComponent
N,N-Dimethylformamide

N,N-Dimethylformamide

Flammable
Health
Irritant
Supplier:   KRAEMER MARTIN
N,N-Dimethylformamide
Supplier:  KRAEMER MARTIN
Certificates
N,N-Dimethylformamide
N,N-Dimethylformamide

2 Options Available Below

Product Details & Documents
Synonyms:  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  • CAS Number:68-12-2
  • Formula:HCON(CH₃)₂
  • MDL Number:MFCD00003284
  • Molecular Weight:73.09 g/mol
  • Merck Index:13,03269
  • Boiling Pt:153 °C (1013 hPa)
  • Melting Pt:–61 °C
  • Density:0.949 g/cm³ (20 °C)
  • Flash Pt:57.5 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-679-5
  • UN:2265
  • ADR:3,III
  • REACH:01-2119475605-32
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 26,80 €
    26,80 €
    1 * 1 L
       26,80 €

  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 350,00 €
    350,00 €
    1 * 5 L
       350,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.