Order Entry
Germany
ContactUsLinkComponent
Propionyl chloride for synthesis, Sigma-Aldrich®
Flammable
Corrosive
Propionyl chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Corrosive
Propionyl chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (rigorous decomposition)
  • Melting Point:
    -94 °C
  • Molar Mass:
    92.52 g/mol
  • Boiling Point:
    80 °C (1013 hPa)
  • Vapor Pressure:
    106 hPa (20 °C)
  • Flash Point:
    6.5 °C
  • Refractive Index:
    1.4032 (20 °C, 589 nm)
  • Explosion Limit:
    3.6 - 11.9 %(V)
  • Density:
    1.06 g/cm3 (20 °C)
  • Ignition Point:
    270 °C

Specification Test Results

Related Information

Frequently Bought Together