Sodium pyruvate
Catalog # A1530.1000
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:110.04 g/mol
Melting Pt:220…230 °C
Density:1.718 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-024-4
CAS Number:113-24-6
Formula:CH₃COCOONa
MDL Number:MFCD00002586