Order Entry
Germany
ContactUsLinkComponent
Pyruvic acid for synthesis, Sigma-Aldrich®
Corrosive
Pyruvic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Pyruvic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Melting Point:
    12 °C
  • Molar Mass:
    88.06 g/mol
  • Flash Point:
    82 - 83 °C
  • Refractive Index:
    1.428 (20 °C, 589 nm)
  • Density:
    1.27 g/cm3 (20 °C)
  • pH:
    1.2 (90 g/l, H2O, 20 °C)
  • Ignition Point:
    305 °C
  • Hill Formula:
    C3H4O3

Specification Test Results

Related Information

Certificates
Search by Lot Number