Acetoacetanilide, Sigma-Aldrich®
SIAL538329-500G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
:
:177.2 g/mol
:13,00059
:129 °C (32 mmHg)
:130…132 °C
:1.26 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:202-996-4
:102-01-2
:C₁₀H₁₁NO₂
:MFCD00008780