7-Tridecanone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:462-18-0
:
Formula:C₁₃H₂₆O
MDL Number:MFCD00009543
:198.35 g/mol
Boiling Pt:264 °C (1013 hPa)
Melting Pt:30…33 °C
Density:0.825 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:207-324-3