
3-Methoxypropiononitrile for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-67-8
:
Formula:C₄H₇NO
MDL Number:MFCD00001958
Boiling Pt:165 °C (1013 hPa)
Melting Pt:–63 °C
Density:0.94 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:85.11 g/mol
EINECS:203-790-7
:GHS07
Specification Test Results
- Solubility:335 g/l (20 °C)
- Melting Point:-63 °C
- Molar Mass:85.11 g/mol
- Boiling Point:164 - 165 °C (1013 hPa)
- Vapor Pressure:9 hPa (50 °C)
- Flash Point:66 °C
- Refractive Index:1.4035 (20 °C, 589 nm)
- Explosion Limit:1.9 - 15.3 %(V)
- Density:0.94 g/cm3 (20 °C)
- pH:6.8 (111 g/l, H2O, 25 °C)
- Ignition Point:390 °C DIN 51794