Order Entry
Germany
ContactUsLinkComponent
Acetonitrile ≥99.93% for biotechnology, Sigma-Aldrich®
Acetonitrile ≥99.93% for biotechnology, Sigma-Aldrich®
Catalog # SIAL494445-100ML
Supplier:  Merck
CAS Number:  
undefined
Acetonitrile ≥99.93% for biotechnology, Sigma-Aldrich®
Catalog # SIAL494445-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
 :M-Clarity™ quality level = MQ200
REACH:01-2119471307-38
 :GHS02
 :GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878