Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trichlorofluoromethane
Trichlorofluoromethane
Trichlorofluoromethane
48541.
:
Merck
:
Trichlorofluoromethane
48541.
:
Merck
:
Pk:
1
Seq:
0001
:
:
137.37 g/mol
:
12,09770
:
23.8 °C (1013 hPa)
:
–111.1 °C
:
1.49 g/cm³ (20 °C)
:
Refrigerator
:
200-892-3
:
75-69-4
:
CCl₃F