Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trichlorofluoromethane
Trichlorofluoromethane, Sigma-Aldrich®
Trichlorofluoromethane, Sigma-Aldrich®
:
Merck
Trichlorofluoromethane, Sigma-Aldrich®
:
Merck
:
75-69-4
:
:
CCl₃F
:
137.37 g/mol
:
12,09770
:
23.8 °C (1013 hPa)
:
–111.1 °C
:
1.49 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ100
:
Refrigerator
:
200-892-3