Order Entry
Germany
ContactUsLinkComponent
Trichlorofluoromethane
Trichlorofluoromethane
Catalog # ABCRAB103650.0100
Supplier:  abcr
CAS Number:  
Trichlorofluoromethane
Catalog # ABCRAB103650.0100
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
 :137.37 g/mol
Merck Index:12,09770
Boiling Pt:23.8 °C (1013 hPa)
Melting Pt:–111.1 °C
Density:1.49 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:200-892-3
CAS Number:75-69-4
Formula:CCl₃F