Trichloroacetic acid (TCA), Sigma-Aldrich®
: Merck
:76-03-9
:
:Cl₃CCOOH
:MFCD00004177
:163.39 g/mol
:13,09700
:196 °C (1013 hPa)
:54…56 °C
:1.63 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:113 °C (closed cup)
:Ambient
:200-927-2
:GHS05
:GHS07
:GHS09