Trichloroacetic acid (TCA), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Merck Index:13,09700
Boiling Pt:196 °C (1013 hPa)
Melting Pt:54…56 °C
Density:1.63 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:113 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-927-2
:GHS05
:GHS07
:GHS09