Order Entry
Germany
ContactUsLinkComponent
Chloroacetic acid for synthesis, Sigma-Aldrich®
Toxic
Corrosive
Environment
Chloroacetic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Toxic
Corrosive
Environment
Chloroacetic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    4210 g/l (20 °C)
  • Melting Point:
    62 °C
  • Molar Mass:
    94.49 g/mol
  • Bulk Density:
    800 kg/m3
  • Boiling Point:
    189 °C (1013 hPa)
  • Vapor Pressure:
    1 hPa (20 °C)
  • Flash Point:
    126 °C
  • Explosion Limit:
    8.0 %(V)
  • Density:
    1.37 g/m3 (65 °C)
  • pH:
     - 1 (800 g/l, H2O, 20 °C)
  • Ignition Point:
    460 °C
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

Specification Test Results

Related Information

Certificates
Search by Lot Number