Dichloroacetonitrile, Sigma-Aldrich®
Catalog # SIAL125954-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:109.94 g/mol
Boiling Pt:112 °C (1013 hPa)
Density:1.374 g/cm³ (11 °C)
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:221-159-4
UN:2920
CAS Number:3018-12-0
Formula:C₂HCl₂N
MDL Number:MFCD00040886