Dichloroacetonitrile
Catalog # EHERCA12321000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:109.94 g/mol
Boiling Pt:112 °C (1013 hPa)
Density:1.374 g/cm³ (11 °C)
EINECS:221-159-4
UN:2920
ADR:8,II
CAS Number:3018-12-0
Formula:C₂HCl₂N
MDL Number:MFCD00040886