Order Entry
Germany
Orders LinkContactUsLinkComponent
1,2,3-Trichloropropane

1,2,3-Trichloropropane

1
Catalog # 47794.
Health
Irritant
Supplier:   Merck
1,2,3-Trichloropropane
Catalog # 47794.
Supplier:  Merck
CAS Number:  96-18-4
1,2,3-Trichloropropane
1,2,3-Trichloropropane

1

CAS Number:  96-18-4
Price
62,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
  • CAS Number:96-18-4
  • Formula:ClH₂CCHClCH₂Cl
  • MDL Number:MFCD00000946
  • Molecular Weight:147.43 g/mol
  • Boiling Pt:156 °C (1013 hPa)
  • Melting Pt:–14.7 °C
  • Density:1.38 g/cm³ (20 °C)
  • Flash Pt:74 °C
  • Storage Temperature:Ambient
  • EINECS:202-486-1
  • UN:2810
  • ADR:6.1,II
Specifications

Specifications

  • Pk:
    1

Specifications