11-Bromo-1-undecanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1611-56-9
:
Formula:HO(CH₂)₁₀CH₂Br
MDL Number:MFCD00004752
:251.21 g/mol
Boiling Pt:350 °C (1013 hPa)
Melting Pt:50 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:216-554-3