Dibromofluoromethane
Catalog # ABCRAB101441.0100
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
About this item
:
:191.83 g/mol
Boiling Pt:64.9 °C (1013 hPa)
Melting Pt:–78 °C
Density:2.48 g/cm³ (20 °C)
Storage Temperature:Refrigerator
UN:1230
ADR:3,II
:GHS02
:GHS06
:GHS08
CAS Number:1868-53-7
Formula:Br₂CFH