
Tetrabromomethane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:558-13-4
:
Formula:Br₄C
MDL Number:MFCD00000117
:331.63 g/mol
Boiling Pt:190 °C (1013 hPa)
Melting Pt:90…92 °C
Density:3.42 g/cm³ (20 °C)
EINECS:209-189-6
ADR:6.1,III
:M-Clarity™ quality level = MQ200
UN:2516
:GHS07
Storage Temperature:Ambient