Potassium sorbate
1
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate
- CAS Number:24634-61-5
- Formula:CH₃CH=CHCH=CHCOOK
- MDL Number:MFCD00016546
- Molecular Weight:150.22 g/mol
- Merck Index:13,07756
- Melting Pt:270 °C
- Density:1.36 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:246-376-1
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:1



