Potassium sorbate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:24634-61-5
:590-00-1
Formula:CH₃CH=CHCH=CHCOOK
MDL Number:MFCD00016546
:150.22 g/mol
Merck Index:13,07756
Melting Pt:270 °C
Density:1.36 g/cm³ (20 °C)
EINECS:246-376-1
:GHS07
Storage Temperature:Ambient