Order Entry
Germany
ContactUsLinkComponent
Potassium sorbate
Potassium sorbate
Catalog # EHERC16972000
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Potassium sorbate
Catalog # EHERC16972000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :150.22 g/mol
Merck Index:13,07756
Melting Pt:270 °C
Density:1.36 g/cm³ (20 °C)
EINECS:246-376-1
 :GHS07
Storage Temperature:Ambient
CAS Number:24634-61-5
 :590-00-1
Formula:CH₃CH=CHCH=CHCOOK
MDL Number:MFCD00016546