Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Potassium sorbate

Glass bottle for solids, 100 mg
Catalog # EDQMP2650000
Irritant
Supplier:   EDQM
Potassium sorbate
Catalog # EDQMP2650000
Supplier:  EDQM
CAS Number:  24634-61-5
Potassium sorbate
Potassium sorbate

Glass bottle for solids, 100 mg

CAS Number:  24634-61-5
Price
270,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate
  • CAS Number:24634-61-5
  • Formula:CH₃CH=CHCH=CHCOOK
  • MDL Number:MFCD00016546
  • Molecular Weight:150.22 g/mol
  • Merck Index:13,07756
  • Melting Pt:270 °C
  • Density:1.36 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:246-376-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.