Order Entry
Germany
ContactUsLinkComponent
1,10-Phenanthroline
1,10-Phenanthroline
Catalog # MOLE62000053-25G
Supplier:  Molekula
CAS Number:  
undefined
1,10-Phenanthroline
Catalog # MOLE62000053-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :180.21 g/mol
Merck Index:12,07356
Boiling Pt:360 °C (1013 hPa)
Melting Pt:117…119 °C
EINECS:200-629-2
UN:2811
ADR:6.1,III
 :GHS06
 :GHS09
CAS Number:66-71-7
Formula:C₁₂H₈N₂
MDL Number:MFCD00011678