1,10-Phenanthroline 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:66-71-7
:
Formula:C₁₂H₈N₂
MDL Number:MFCD00011678
:180.21 g/mol
Merck Index:12,07356
Boiling Pt:360 °C (1013 hPa)
Melting Pt:117…120 °C
Storage Temperature:Ambient
EINECS:200-629-2
UN:2811
ADR:6.1,III
:GHS06
:GHS09
Specification Test Results
- Appearance:White to light yellow to light pink Powder
- Infrared spectrum:Conforms
- Melting point:114 to 120 °C
- Sulfated ash:≤0.1 %
- Titration with HClO4:≥99.0 %