Order Entry
Germany
Orders LinkContactUsLinkComponent
D-(+)-Biotin

D-(+)-Biotin

Marketsource
Supplier:   DIANOVA
D-(+)-Biotin
Supplier:  DIANOVA
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Merck Index:13,01231
  • Melting Pt:232…233 °C
  • Density:1.41 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:200-399-3
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 20 µG
    Log in for availability
    Log in for availability
    1 * 20 µG
    1 * 20 µG

    Specifications:
    Marketsource
    Marketsource
  • 100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.