Order Entry
Germany
Orders LinkContactUsLinkComponent
D-(+)-Biotin 98%

D-(+)-Biotin 98%

D-(+)-Biotin 98%
Certificates
D-(+)-Biotin 98%

4 Options Available Below

Product Details & Documents
Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Merck Index:13,01231
  • Melting Pt:231…233 °C (dec.)
  • Density:1,41 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:200-399-3

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 % (105°C)
  • Sulfated ash:
    ≤0.1 %
  • Appearance of solution:
    (2% soln. in 0.1 N NaOH) clear, colorless
  • Specific optical rotation:
    +89° to +93° (20°C, 589 nm) (c=2, 0.1 N NaOH) (on dried substance)
  • Titration with NaOH:
    ≥97.5 % (On dried substance)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 82,00 €
    82,00 €
    1 * 500 mg
       82,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 134,00 €
    134,00 €
    1 * 1 g
       134,00 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 457,00 €
    457,00 €
    1 * 5 g
       457,00 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 800,00 €
    1 800,00 €
    1 * 25 g
       1 800,00 €