Order Entry
Germany
ContactUsLinkComponent
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid for synthesis
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid for synthesis
Catalog # 8.24449.0250
Supplier:  Merck
CAS Number:  
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid for synthesis
Catalog # 8.24449.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    177 - 180 °C
  • Molar Mass:
    172.18 g/mol
  • Synonyms:
    (1R,2R)-(-)-Hexahydrophthalic acid
  • Hill Formula:
    C8H12O4

Specification Test Results

About this item

 :
 :172.18 g/mol
 :M-Clarity™ quality level = MQ200
CAS Number:46022-05-3
Formula:C₈H₁₂O₄
MDL Number:MFCD00062973