
(R)-(+)-2-Phenyl-1-propanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19141-40-3
:
Formula:C₉H₁₂O
MDL Number:MFCD00145206
:136.19 g/mol
Boiling Pt:220 °C
Density:0.975
Flash Pt:108 °C (226 °F)
:GHS07
Storage Temperature:Ambient