L(-)-Tyrosine
Supplier: BIOMOL RESEARCH LABORATORIES
Certificates
About this item
CAS Number:60-18-4
:
Formula:C₉H₁₁NO₃
MDL Number:MFCD00002606
:181.19 g/mol
Merck Index:13,09907
Boiling Pt:351 °C (1013 hPa)
Melting Pt:285 °C
Density:1.41 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-460-4