
(R)-(-)-Hydratropic acid 99%
ACRO264662500
:
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
- Appearance:Colorless to white to yellow to light brown Crystals or needles or low melting solid or liquid
- Infrared spectrum:Conforms
- GC:≥98.5 %
- Specific optical rotation:-75° to -82° (20°C, 589 nm) (c=1.5, EtOH)
:
:150.18 g/mol
:115 °C (1 mmHg)
:29…32 °C
:1.1 g/cm³
:>110 °C
:GHS07
:Refrigerator
:7782-26-5
:C₉H₁₀O₂
:MFCD00063140