Order Entry
Germany
ContactUsLinkComponent
L-Tryptophan, EMPROVE® EXPERT Ph. Eur., BP, JPC suitable for use as excipient, SAFC®
L-Tryptophan, EMPROVE® EXPERT Ph. Eur., BP, JPC suitable for use as excipient, SAFC®
Catalog # 1.08396.1000
Supplier:  Merck
CAS Number:  
L-Tryptophan, EMPROVE® EXPERT Ph. Eur., BP, JPC suitable for use as excipient, SAFC®
Catalog # 1.08396.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :204.23 g/mol
Merck Index:12,09929
Melting Pt:290 °C
Density:1.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-795-6
CAS Number:73-22-3
Formula:C₁₁H₁₂N₂O₂
MDL Number:MFCD00064340