Order Entry
Germany
ContactUsLinkComponent
(S)-(+)-2-Amino-3-methyl-1-butanol
(S)-(+)-2-Amino-3-methyl-1-butanol
Catalog # MOLE19625420-1G
Supplier:  Molekula
CAS Number:  
(S)-(+)-2-Amino-3-methyl-1-butanol
Catalog # MOLE19625420-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :103.16 g/mol
Boiling Pt:187 °C (1013 hPa)
Melting Pt:31…32 °C
Density:0.92 g/cm³ (20 °C)
EINECS:217-975-5
CAS Number:2026-48-4
Formula:C₅H₁₃NO
MDL Number:MFCD00064296