L(+)-Leucine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:61-90-5
:
Formula:C₆H₁₃NO₂
MDL Number:MFCD00002617
:131.17 g/mol
Merck Index:14,05451
Melting Pt:300 °C
Density:1.165 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-522-0