Order Entry
Germany
ContactUsLinkComponent
L(+)-Leucine
L(+)-Leucine
Catalog # MOLEM72696989
Supplier:  Molekula
CAS Number:  
L(+)-Leucine
Catalog # MOLEM72696989
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :131.17 g/mol
Merck Index:14,05451
Melting Pt:300 °C
Density:1.165 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-522-0
CAS Number:61-90-5
Formula:C₆H₁₃NO₂
MDL Number:MFCD00002617