Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(+)-Leucine
L(+)-Leucine
L(+)-Leucine
ABCRAB117225.0002
:
abcr
:
L(+)-Leucine
ABCRAB117225.0002
:
abcr
:
Pack type:
Glass bottle for solids
Pk:
2,5 kg
:
:
131.17 g/mol
:
14,05451
:
300 °C
:
1.165 g/cm³ (20 °C)
:
Ambient
:
200-522-0
:
61-90-5
:
C₆H₁₃NO₂
:
MFCD00002617