Order Entry
Germany
Orders LinkContactUsLinkComponent
 

L(+)-Valine

L(+)-Valine
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  H-Val-OH, Valine, (S)-(+)-Valine
  • CAS Number:72-18-4
  • Formula:C₅H₁₁NO₂
  • MDL Number:MFCD00064220
  • Molecular Weight:117.15 g/mol
  • Merck Index:13,09458
  • Boiling Pt:217 °C (1013 hPa)
  • Melting Pt:86 °C
  • Density:1.284 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-773-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g