D-(-)-Valine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:640-68-6
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD00064219
Boiling Pt:217 °C (1013 hPa)
Melting Pt:86 °C
:M-Clarity™ quality level = MQ200
:117.15 g/mol
EINECS:211-368-9