Order Entry
Germany
Orders LinkContactUsLinkComponent
 

D-(-)-Sorbitol

Glass bottle for solids, 500 g
Catalog # SERA39785.01
Marketsource
Supplier:   Serva
D-(-)-Sorbitol
Catalog # SERA39785.01
Supplier:  Serva
Marketsource
CAS Number:  50-70-4
D-(-)-Sorbitol
D-(-)-Sorbitol

Glass bottle for solids, 500 g

CAS Number:  50-70-4
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,08797
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:94…98 °C
  • Density:1.524 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications