(R)-(-)-2-Butanol
Catalog # MOLEM24349831
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:74.12 g/mol
Merck Index:13,01540
Boiling Pt:99 °C (1013 hPa)
Melting Pt:–114 °C
Density:0.807 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:238-967-8
UN:1120
ADR:3,III
:GHS02
:GHS07
CAS Number:14898-79-4
Formula:C₄H₁₀O
MDL Number:MFCD00064280