(R)-(-)-1-Methoxy-2-propanol, Sigma-Aldrich®
Catalog # SIAL77913-5ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:90.12 g/mol
Storage Temperature:Ambient
UN:3092
ADR:3,III
CAS Number:4984-22-9
:49840-22-9
Formula:C₄H₁₀O₂
MDL Number:MFCD01632587