Order Entry
Germany
ContactUsLinkComponent
(R)-(-)-1-Methoxy-2-propanol
(R)-(-)-1-Methoxy-2-propanol
Catalog # MOLEM10995842
Supplier:  Molekula
CAS Number:  
(R)-(-)-1-Methoxy-2-propanol
Catalog # MOLEM10995842
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :90.12 g/mol
Storage Temperature:Ambient
UN:3092
ADR:3,III
CAS Number:4984-22-9
 :49840-22-9
Formula:C₄H₁₀O₂
MDL Number:MFCD01632587