(S)-(+)-sec-Butylamine, Sigma-Aldrich®
Catalog # SIAL296651-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:73.14 g/mol
Merck Index:12,01579
Boiling Pt:61 °C (1013 hPa)
Melting Pt:–104 °C
Density:0.724 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-164-7
UN:2733
ADR:3,II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS05
:GHS07
:GHS09
CAS Number:513-49-5
Formula:C₄H₁₁N
MDL Number:MFCD00064417