Order Entry
Germany
ContactUsLinkComponent
L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    99.0 - 102.0 %
  • pH (25°C; 5 %):
    7.0 - 8.5
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • NH4 (Ammonium):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • PO4 (Phosphate):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Ca (Calcium):
    Max. 40 ppm
  • Cu (Copper):
    Max. 5 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Pb (Lead):
    Max. 5 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results