Supelco® Organic Reference Standard, Benzo[A] Pyrene Solution
31306-10ML
: Merck
:
- Pack type:Glass bottle
- Pk:10 mL
- Standards:Analytical standard
- CAS No.:50-32-8
- Description:Benzo[a]pyrene solution
- Storage conditions:2...8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H350 - H410
- Shipping conditions:STD
- MDL No.:MFCD00003602
- M-Clarity quality level:MQ100
- Formula:C₂₀H₁₂
- Molecular weight:252,31
- Flash point:–18 °C
- GHS signal: GHS02, GHS08, GHS07, GHS09
: P202 - P210 - P273 - P301 + P310 - P303 + P361 + P353 - P331