Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, 2,2',3,3',4,5,5',6,6'-Nonachlorobiphenyl (PCB No. 208) 10 µg/ml in isooctane, Dr Ehrenstorfer
Organic reference standard, 2,2',3,3',4,5,5',6,6'-Nonachlorobiphenyl (PCB No. 208) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20020800IO
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2,2',3,3',4,5,5',6,6'-Nonachlorobiphenyl (PCB No. 208) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20020800IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    52663-77-1
  • Description:
    2,2',3,3',4,5,5',6,6'-Nonachlorobiphenyl (PCB No. 208) 10 µg/ml in isooctane
  • Formula:
    C₁₂HCl₉
  • Description:
    2,2',3,3',4,5,5',6,6'-Nonachlorobiphenyl (PCB No. 208)

Specifications

About this item