Organic reference standard, 2,3,5,6-Tetrachlorobiphenyl (PCB No. 65) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-65
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:33284-54-7
- Description:2,3,5,6-Tetrachlorobiphenyl (PCB No. 65) 35 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,3,5,6-Tetrachlorobiphenyl (PCB No. 65)