Organic reference standard, 2,3,4,5-Tetrachlorobiphenyl (PCB No. 61) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-61
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:33284-53-6
- Description:2,3,4,5-Tetrachlorobiphenyl (PCB No. 61) 35 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,3,4,5-Tetrachlorobiphenyl (PCB No. 61)